ENA360887694 Enamine

methyl 5-amino-2,3-difluorobenzoate

  • MDL number MFCD16875975
  • Cas Number : 1227269-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99468

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99468
Availability details for
Quantity
  • Availability
  • Add to Cart