ENA360887701 Enamine

3-(4-nitrophenyl)-1,2,4-oxadiazol-5-amine

  • MDL number MFCD16764849
  • Cas Number : 23275-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 287-288
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-99652

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-99652
Availability details for
Quantity
  • Availability
  • Add to Cart