ENA360887703 Enamine

1-(1,3,5-trimethyl-1H-pyrazole-4-amido)cyclopentane-1-carboxylic acid

  • MDL number MFCD12569326
  • Cas Number : 1152866-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 235...237
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-99496

Safety Information

msds https://www.enaminestore.com/catalog/EN300-99496
Availability details for
Quantity
  • Availability
  • Add to Cart