ENA369381410 Enamine

ethyl 2-chloro-4-methylquinoline-3-carboxylate

  • MDL number MFCD22056321
  • Cas Number : 251902-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109...111
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100010

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100010
Availability details for
Quantity
  • Availability
  • Add to Cart