ENA369381427 Enamine

5-(benzyloxy)-2-(hydroxymethyl)-1-methyl-1,4-dihydropyridin-4-one

  • MDL number MFCD22056356
  • Cas Number : 89539-51-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100427

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100427
Availability details for
Quantity
  • Availability
  • Add to Cart