ENA369381439 Enamine

(3S)-3-amino-3-(4-fluorophenyl)propanamide hydrochloride

  • MDL number MFCD22056361
  • Cas Number : 1384435-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100514
Availability details for
Quantity
  • Availability
  • Add to Cart