ENA369381442 Enamine

methyl 5-[(dimethylcarbamoyl)sulfanyl]-4-methoxy-2-nitrobenzoate

  • MDL number MFCD22056362
  • Cas Number : 1384428-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100525

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100525
Availability details for
Quantity
  • Availability
  • Add to Cart