ENA369381446 Enamine

N2,N2,N3-trimethylpyridine-2,3-diamine hydrochloride

  • MDL number MFCD22056370
  • Cas Number : 1384427-84-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100717

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100717
Availability details for
Quantity
  • Availability
  • Add to Cart