ENA369381449 Enamine

1-amino-N,N-dimethylcyclopentane-1-carboxamide hydrochloride

  • MDL number MFCD22056375
  • Cas Number : 1384427-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100772

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100772
Availability details for
Quantity
  • Availability
  • Add to Cart