ENA369381458 Enamine

5-(benzyloxy)-2-(chloromethyl)-1-methyl-1,4-dihydropyridin-4-one hydrochloride

  • MDL number MFCD22056384
  • Cas Number : 481726-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100901
Availability details for
Quantity
  • Availability
  • Add to Cart