ENA369381460 Enamine

2-(2-methylphenoxy)butan-1-amine hydrochloride

  • MDL number MFCD22056386
  • Cas Number : 1384428-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100946

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100946
Availability details for
Quantity
  • Availability
  • Add to Cart