ENA369381462 Enamine

2-bromo-5-chloro-4-methoxybenzene-1-sulfonyl chloride

  • MDL number MFCD22196381
  • Cas Number : 868406-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-100971

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100971
Availability details for
Quantity
  • Availability
  • Add to Cart