ENA369381494 Enamine

N-[(2-ethoxyphenyl)methyl]-2H-indazol-6-amine

  • MDL number MFCD22056427
  • Cas Number : 415969-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101388

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101388
Availability details for
Quantity
  • Availability
  • Add to Cart