ENA369381495 Enamine

1-methyl-5-(trichloroacetyl)-1H-pyrrole-3-carbonitrile

  • MDL number MFCD22196401
  • Cas Number : 135250-39-4
  • Molecular Weight : 251.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101421

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101421
Availability details for
Quantity
  • Availability
  • Add to Cart