ENA369381497 Enamine

1-(2-chloroethyl)-4-phenyl-1H-pyrazole hydrochloride

  • MDL number MFCD22056428
  • Cas Number : 1384430-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101487

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101487
Availability details for
Quantity
  • Availability
  • Add to Cart