ENA369381502 Enamine

2-(4-propylpiperidin-1-yl)cyclohexan-1-one hydrochloride

  • MDL number MFCD22196410
  • Cas Number : 1384430-26-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101542

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101542
Availability details for
Quantity
  • Availability
  • Add to Cart