ENA369381508 Enamine

6,7-dinitro-2,3-dihydro-1,4-benzodioxine-5-carboxylic acid

  • MDL number MFCD22196414
  • Cas Number : 66411-42-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101720

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101720
Availability details for
Quantity
  • Availability
  • Add to Cart