ENA369381509 Enamine

4-[2-(morpholin-4-yl)ethyl]-1,7lambda6-dithia-4-azaspiro[4.4]nonane-3,7,7-trione

  • MDL number MFCD22196416
  • Cas Number : 1384428-57-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101733

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101733
Availability details for
Quantity
  • Availability
  • Add to Cart