ENA369381511 Enamine

2-[(1,1-dioxo-2,5-dihydro-1lambda6-thiophen-3-yl)sulfanyl]-N-phenylacetamide

  • MDL number MFCD22196421
  • Cas Number : 1123204-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101784

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101784
Availability details for
Quantity
  • Availability
  • Add to Cart