ENA369381515 Enamine

(cyclobutylmethyl)(2,2-difluoroethyl)amine hydrochloride

  • MDL number MFCD22196428
  • Cas Number : 1384427-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101815

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-101815
Availability details for
Quantity
  • Availability
  • Add to Cart