ENA369381516 Enamine

1-[5-(4-chlorophenyl)-1,2,4-oxadiazol-3-yl]ethan-1-amine hydrochloride

  • MDL number MFCD22196429
  • Cas Number : 1384429-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 232...234
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101846

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101846
Availability details for
Quantity
  • Availability
  • Add to Cart