ENA369381523 Enamine

but-2-yn-1-amine hydrochloride

  • MDL number MFCD22196444
  • Cas Number : 50329-23-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-101997

Safety Information

msds https://www.enaminestore.com/catalog/EN300-101997
Availability details for
Quantity
  • Availability
  • Add to Cart