ENA369381524 Enamine

N-[1-(2-fluorophenyl)ethyl]cyclopentanamine hydrochloride

  • MDL number MFCD22196448
  • Cas Number : 1384430-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102045

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102045
Availability details for
Quantity
  • Availability
  • Add to Cart