ENA369381528 Enamine

5-fluoro-2-(2,2,2-trifluoroethoxy)aniline hydrochloride

  • MDL number MFCD22196452
  • Cas Number : 1384429-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189...191
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102100

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102100
Availability details for
Quantity
  • Availability
  • Add to Cart