ENA369381529 Enamine

5-methyl-2-phenoxyaniline hydrochloride

  • MDL number MFCD22196454
  • Cas Number : 166100-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102125

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102125
Availability details for
Quantity
  • Availability
  • Add to Cart