ENA369381534 Enamine

(2,3,5,6-tetrafluorophenyl)methanamine hydrochloride

  • MDL number MFCD22196463
  • Cas Number : 158388-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 279...281
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102269

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102269
Availability details for
Quantity
  • Availability
  • Add to Cart