ENA369381536 Enamine

2-chloro-3-(pyrrolidin-1-ylmethyl)pyridine hydrochloride

  • MDL number MFCD22196468
  • Cas Number : 1384429-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 214...216
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102329

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102329
Availability details for
Quantity
  • Availability
  • Add to Cart