ENA369381537 Enamine

3-methanesulfonylcyclohexyl methanesulfonate

  • MDL number MFCD22196469
  • Cas Number : 1384428-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102349

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102349
Availability details for
Quantity
  • Availability
  • Add to Cart