ENA369381539 Enamine

(3R)-3-amino-3-(4-fluorophenyl)propanamide hydrochloride

  • MDL number MFCD22196472
  • Cas Number : 1384424-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102366

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-102366
Availability details for
Quantity
  • Availability
  • Add to Cart