ENA369381546 Enamine

1,4-dichloronaphthalene

  • MDL number MFCD00152414
  • Cas Number : 1825-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-68
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-100379

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100379
Availability details for
Quantity
  • Availability
  • Add to Cart