ENA369381551 Enamine

6-(4-bromo-1H-pyrazol-1-yl)pyridazine-3-carboxylic acid

  • MDL number MFCD12779159
  • Cas Number : 1183884-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100392

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100392
Availability details for
Quantity
  • Availability
  • Add to Cart