ENA369381554 Enamine

2-(3-chlorophenyl)-3-methylbutanoic acid

  • MDL number MFCD11936727
  • Cas Number : 55776-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100395

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100395
Availability details for
Quantity
  • Availability
  • Add to Cart