ENA369381565 Enamine

butyl({[1-(4-methoxyphenyl)-2,5-dimethyl-1H-pyrrol-3-yl]methyl})amine

  • MDL number MFCD22196362
  • Cas Number : 1243598-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-100408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100408
Availability details for
Quantity
  • Availability
  • Add to Cart