ENA369381570 Enamine

1-[4-(dimethylamino)-2-hydroxyphenyl]ethan-1-one

  • MDL number MFCD11870193
  • Cas Number : 107070-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120...122
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100023

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100023
Availability details for
Quantity
  • Availability
  • Add to Cart