ENA369381574 Enamine

2-(3-methylphenoxy)butanoic acid

  • MDL number MFCD03422243
  • Cas Number : 113104-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 72...74
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100419

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100419
Availability details for
Quantity
  • Availability
  • Add to Cart