ENA369381586 Enamine

1-(3,4-dichlorophenyl)-3-methyl-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD00849838
  • Cas Number : 13124-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100433

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100433
Availability details for
Quantity
  • Availability
  • Add to Cart