ENA369381588 Enamine

4-amino-3-methoxy-N,N-dimethylbenzamide hydrochloride

  • MDL number MFCD22056357
  • Cas Number : 1376330-43-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100435

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100435
Availability details for
Quantity
  • Availability
  • Add to Cart