ENA369381592 Enamine

2-(bromomethyl)-4-methoxy-1-nitrobenzene

  • MDL number MFCD08234906
  • Cas Number : 67567-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100441

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100441
Availability details for
Quantity
  • Availability
  • Add to Cart