ENA369381594 Enamine

N-[1-(4-chlorophenyl)ethyl]aniline

  • MDL number MFCD11138684
  • Cas Number : 92252-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100030

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100030
Availability details for
Quantity
  • Availability
  • Add to Cart