ENA369381597 Enamine

(1R)-6-fluoro-1,2,3,4-tetrahydronaphthalen-1-amine hydrochloride

  • MDL number MFCD22056325
  • Cas Number : 1373232-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100032

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100032
Availability details for
Quantity
  • Availability
  • Add to Cart