ENA369381603 Enamine

3-amino-1,1-difluoropropan-2-ol

  • MDL number MFCD16805790
  • Cas Number : 433-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100454

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100454
Availability details for
Quantity
  • Availability
  • Add to Cart