ENA369381604 Enamine

4-propanoyl-1,2,3,4-tetrahydroquinoxalin-2-one

  • MDL number MFCD03163555
  • Cas Number : 923692-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100052

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100052
Availability details for
Quantity
  • Availability
  • Add to Cart