ENA369381610 Enamine

3-methyl-5-phenylcyclohex-2-en-1-one

  • MDL number MFCD22056359
  • Cas Number : 5337-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100474

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100474
Availability details for
Quantity
  • Availability
  • Add to Cart