ENA369381611 Enamine

N-butylbutanamide

  • MDL number MFCD03367403
  • Cas Number : 10264-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100475

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100475
Availability details for
Quantity
  • Availability
  • Add to Cart