ENA369381613 Enamine

1-methylcyclobutane-1-carboxylic acid

  • MDL number MFCD15527494
  • Cas Number : 32936-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100478

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100478
Availability details for
Quantity
  • Availability
  • Add to Cart