ENA369381625 Enamine

ethyl 3-(benzylamino)-2-methylpropanoate

  • MDL number MFCD00100898
  • Cas Number : 99985-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100498

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100498
Availability details for
Quantity
  • Availability
  • Add to Cart