ENA369381626 Enamine

4-bromo-1-{[3-(trifluoromethyl)phenyl]methyl}-1H-pyrazole

  • MDL number MFCD02828766
  • Cas Number : 1006538-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100053

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100053
Availability details for
Quantity
  • Availability
  • Add to Cart