ENA369381631 Enamine

5-chloro-2-(2-methoxyethoxy)benzoic acid

  • MDL number MFCD02668674
  • Cas Number : 62176-27-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100054
Availability details for
Quantity
  • Availability
  • Add to Cart