ENA369381632 Enamine

1-(2-phenoxyethyl)cyclopropan-1-ol

  • MDL number MFCD16840893
  • Cas Number : 1247788-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57...59
Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-100056

Safety Information

msds https://www.enaminestore.com/catalog/EN300-100056
Availability details for
Quantity
  • Availability
  • Add to Cart