ENA369381633 Enamine

[3-(difluoromethoxy)phenyl]methanethiol

  • MDL number MFCD12026282
  • Cas Number : 1208074-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-100517

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-100517
Availability details for
Quantity
  • Availability
  • Add to Cart